Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIF3A Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA579362
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue.
Cell growth and viability is compromised by oxygen deprivation (hypoxia). Hypoxia-inducible factors, including HIF-1 alpha, HIF-1 beta (also designated Arnt 1), EPAS-1 (also designated HIF-2 alpha) and HIF-3 alpha, induce glycolysis, erythropoiesis and angiogenesis in order to restore oxygen homeostasis. Hypoxia-inducible factors are members of the Per-Arnt-Sim (PAS) domain transcription factor family. In response to hypoxia, HIF-1 alpha is upregulated and forms a heterodimer with Arnt 1 to form the HIF-1 complex. The HIF-1 complex recognizes and binds to the hypoxia responsive element (HRE) of hypoxia-inducible genes, thereby activating transcription.
Specifications
| HIF3A | |
| Polyclonal | |
| Unconjugated | |
| HIF3A | |
| Basic-helix-loop-helix-PAS protein MOP7; BHLHE17; Class E basic helix-loop-helix protein 17; HIF-3 alpha; HIF3 alpha 1; HIF3A; HIF-3A; HIF-3-alpha; HIF3-alpha; HIF3-alpha-1; hypoxia inducible factor 3 alpha; hypoxia inducible factor 3 alpha subunit; hypoxia inducible factor 3, alpha subunit; hypoxia inducible factor 3a; hypoxia inducible factor three alpha; hypoxia-inducible factor 3 alpha; hypoxia-inducible factor 3-alpha; Inhibitory PAS domain protein; IPAS; Member of PAS protein 7; Mop7; Neonatal and embryonic PAS protein; NEPAS; PAS domain-containing protein 7; PASD7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64344, 64345 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q9JHS2, Q9Y2N7 | |
| HIF3A | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human HIF3 (497-514aa DDDFQLNASEQLPRAYHR). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction