Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IFN-gamma DuoSet ELISA

Catalog No. dy285b05
Click to view available options
Sufficient For:
15 x 96-well Plates
5 x 96-well Plates
This item is not returnable. View return policy
This item is not returnable. View return policy

Human IFN-gamma DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human IFN-gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IFN-gamma.

Specifications

Assay Range 9.38 - 600pg/mL
Immunoassay Kit Format 96-well Plate
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum
Sufficient For 5 x 96-well Plates
Target Species Human
Gene ID (Entrez) 3458 (Human); 15978 (Mouse); 25712 (Rat); 396991 (Porcine); 281237 (Bovine); 403801 (Canine); 493965 (Feline)
Kit Contents 96 well Micro Plates, PBS, Plate Sealers, Reagent Diluents Concentrate 2, Stop Solution, Substrate Solution, Wash Buffer
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Target Interferon gamma

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.