Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIG1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS9066R

Description
Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.Specifications
HIG1 | |
Polyclonal | |
Unconjugated | |
HIGD1A | |
2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; HIG1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; Hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a | |
Rabbit | |
Protein A | |
RUO | |
25994 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y241 | |
HIGD1A | |
KLH conjugated synthetic peptide derived from human HIG1/HIGD1A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction