Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIGD1A Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP281721C
This item is not returnable.
View return policy
Description
HIGD1A Polyclonal antibody specifically detects HIGD1A in Human samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
HIGD1A | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
DyLight 650 | |
7420700H20Rik, DKFZP564K247, HIG1 domain family member 1A, HIG1 domain family, member 1A, HIG1 hypoxia inducible domain family, member 1A, HIG1DKFZp564K247, hypoxia inducible gene 1,2210020B17Rik, Hypoxia-inducible gene 1 protein | |
HIGD1A antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human HIGD1A. The immunogen is located within the first 50 amino acids of HIGD1A. Amino Acid Squence: SLPSYEEDQGSKLIRKAKE | |
0.1 mL | |
Cell Biology | |
25994 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction