Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100262 Supplier Invitrogen™ Supplier No. PA5100262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HIGD1A.

Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD1A
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HIGD1A
Gene Accession No. Q8VH49, Q9JLR9, Q9Y241
Gene Alias 2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; Hig1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a
Gene Symbols HIGD1A
Host Species Rabbit
Immunogen A synthesized peptide derived from human HIGD1A(Accession Q9Y241), corresponding to amino acid residues F29-M79.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140937, 25994, 56295
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.