Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD1A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572706
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572706 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572706 Supplier Invitrogen™ Supplier No. PA572706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIGD1A Polyclonal Antibody for Western Blot, ICC/IF, IHC, ELISA

Proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. May play a role in the assembly of respiratory supercomplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD1A
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HIGD1A
Gene Accession No. Q9Y241
Gene Alias 2210020B17Rik; 7420700H20Rik; AI303276; AW049839; DKFZP564K247; Hig1; HIG1 domain family member 1A, mitochondrial; HIG1 domain family, member 1A; HIG1 hypoxia inducible domain family member 1A; HIG1 hypoxia inducible domain family, member 1A; Higd1a; HIMP1; HSPC010; hypoxia induced gene 1; hypoxia inducible gene 1; hypoxia-inducible gene 1 protein; RCF1 homolog A; RCF1a
Gene Symbols HIGD1A
Host Species Rabbit
Immunogen HIG1 antibody for a 19 amino acid synthetic peptide near the amino terminus of human HIG1. The immunogen is within the first 50 amino acids of HIG1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.