Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIGD1A Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7202135 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB093216 20 μL
NB093215 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB093216 Supplier Novus Biologicals Supplier No. NBP30393020UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIGD1A Polyclonal specifically detects HIGD1A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HIGD1A
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias 7420700H20Rik, DKFZP564K247, HIG1 domain family member 1A, HIG1 domain family, member 1A, HIG1 hypoxia inducible domain family, member 1A, HIG1DKFZp564K247, hypoxia inducible gene 1,2210020B17Rik, Hypoxia-inducible gene 1 protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-93 of human HIGD1A (NP_054775.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25994
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.