Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIGD1B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983817
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3817 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3817 Supplier Bioss Supplier No. BS15489R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen HIGD1B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HIGD1B
Gene Accession No. Q9P298
Gene Alias CLST11240; CLST11240-15; HIG1 domain family member 1B; HIG1 hypoxia inducible domain family member 1B; HIG1 hypoxia inducible domain family, member 1B; HIGD1B; Protein CLST 11240
Gene Symbols HIGD1B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human HIGD1B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51751
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.