Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD1C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA549185
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA549185 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA549185 Supplier Invitrogen™ Supplier No. PA549185
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIGD1C Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD1C
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Higd1c
Gene Accession No. A8MV81, Q76I25
Gene Alias Gm921; HIG1 domain family member 1C; HIG1 domain family, member 1C; HIG1 hypoxia inducible domain family member 1C; HIG1 hypoxia inducible domain family, member 1C; HIG1-4; Higd1c; UBIE; UBIE2; YGHL1 homolog; Yghl1-4
Gene Symbols Higd1c
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 69-97 amino acids from the C-terminal region of human HIGD1C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 380975, 613227
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.