Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD2A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572707
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572707 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572707 Supplier Invitrogen™ Supplier No. PA572707
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIGD2A Polyclonal Antibody for Western Blot, ELISA

HIGD2A is a proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. It may be involved in cytochrome c oxidase activity. It may play a role in the assembly of respiratory supercomplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD2A
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene HIGD2A
Gene Accession No. Q9BW72, Q9CQJ1
Gene Alias 2010110M21Rik; AA690237; HIG1 domain family member 2A; HIG1 domain family member 2A, mitochondrial; HIG1 domain family, member 2A; HIG1 hypoxia inducible domain family member 2A; HIG1 hypoxia inducible domain family, member 2A; Higd2a; RCF1 homolog B; RCF1b
Gene Symbols HIGD2A
Host Species Rabbit
Immunogen HIG2 antibody for a 16 amino acid synthetic peptide near the carboxy terminus of human HIG2. The immunogen is within the last 50 amino acids of HIG2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192286, 67044
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.