Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIGD2B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582657 Supplier Invitrogen™ Supplier No. PA582657
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATLGFVTPE APFESSKPPI FEGLSPTVYS NPEGFKEKFL RKTRENP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 79%.

HIGD2B is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD2B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HIGD2B
Gene Accession No. Q4VC39
Gene Alias HIG1 hypoxia inducible domain family member 2B; HIGD2B; HIGD2BP
Gene Symbols HIGD2B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HIGD2B. Recombinant protein control fragment (Product #RP-91011).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 123346
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.