Learn More
Mopec™ High Capacity Section Dryer Timer (5-8)
Click to view available options
Product Type:
Dryer Timer
Description
- Fan-assisted heating dramatically reduces drying time
- Designed to take up minimal bench-top space
- Digital temperature control
- Rapid heating system ensures system is ready to use in less time
- Anti-microbial coating to help prevent bacteria and virus growth
- Timer Units (BK701-T1 and BK701-T2) allow monitoring of four slide racks each and ensures that each rack has the correct amount of time in the dryer
Specifications
Specifications
Product Type | Dryer Timer |
For Use With (Equipment) | High Capacity Section Dryer |