Description
- Protects against dusts and pollens
- Soft, non-irritating inner facepiece
- Adjustable nosepiece
- Heavy-duty, latex-free headstraps
- Dust masks are Not NIOSH approved
Specifications
Specifications
| Product Type | General Purpose Face Mask |
| Color | White |
| Fastening Type | Headband |
| Style | Cone |
| Model | AM175 |
| Packaging Quantity | 10 Packs of 50 |