Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Microporous Lab Coats

Catalog No. 19167921 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-921 Large
19-167-922 X-Large
19-167-923 2X-Large
19-167-924 3X-Large
19-167-920 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-167-921 Supplier High Five™ Supplier No. EW20208L
This item is not returnable. View return policy

Breathability with durability

  • Cloth-like, microporous material
  • General purpose protection from dirt, grime, splashes and spills
  • Economical and lightweight
  • Low linting and antistatic
  • Elastic wrists, five-snap closure and two pockets
  • White

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Large
Gender Unisex
Material Microporous Film
Color White
Closure Type Snaps
No. of Pockets 2
For Use With (Application) General purpose protection from dirt, grime, splashes, and spills
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.