Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell MICROFLEX™ N25 Nitrile Gloves

Catalog No. 19167171 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-167-171 Small
19-167-172 Medium
19-167-173 Large
19-167-174 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-167-171 Supplier Ansell Supplier No. N251
This item is not returnable. View return policy

Durable industrial nitrile glove designed for tough tasks requiring lasting performance including life sciences, automotive, machinery and food processing.

  • Nonsterile
  • Ambidextrous
  • Palm thickness: 5 mil
  • Beaded cuff
  • Textured fingers
  • Length: 9.5 in.
  • Meets FDA food contact requirements
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance FDA
AQL 4
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 5 mil
Tensile Strength 20.5
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.