Learn More
Ansell MICROFLEX™ N58 Nitrile Gloves - Class 100
Catalog No. 19256002 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-256-002 Small
19-256-003 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-256-002 Supplier Ansell Supplier No. N581
This item is not returnable. View return policy
N58 nitrile gloves are 100% latex-Free and are compatible with Class 100/ISO 5 Cleanrooms.
  • ISO Class 5 (Class 100) nitrile glove
  • Odor-free
  • Offers an unequaled fit and feel compared to conventional exam gloves
  • Unique synthetic formulation reduces hand fatigue
  • Exceptional puncture resistance and chemical resistance compared to latex or vinyl gloves
  • Powder free
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance AQL 1.5, ISO9001 QMS
Color White
Disposable Single-use
Material Nitrile
Powdered Powder Free
Size Small
Sterility Non-sterile
Thickness (English) 4.5 mil
Model N581
Tensile Strength 20.5
Length (English) 12 in.
Length (Metric) 30.5 cm