Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ SMS Disposable Lab Coats

Catalog No. 19167927 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Purple
White
Size:
2X-Large
3X-Large
4X-Large
6X-Large
Large
Large/X-Large
Medium
Small
X-Large
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-167-927 White Large/X-Large
19-156-225 Purple Medium
19-156-226 Purple Large
19-156-228 Purple 2X-Large
19-167-630 Blue Large
19-167-631 Blue X-Large
19-167-632 Blue 2X-Large
19-167-633 Blue 3X-Large
19-167-925 White Medium
19-167-926 White Large
19-167-928 White 2X-Large
19-167-929 White 3X-Large
19-167-032 White 4X-Large
19-167-040 Blue Medium
19-167-802 Blue 6X-Large
19-167-820 White Small
19-156-229 Purple 3X-Large
19-167-803 Blue 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Catalog No. 19-167-927 Supplier High Five™ Supplier No. EW15204XL

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Breathable for all day comfort

  • Protect against dry particulates and water-based liquids
  • Knit collar and cuffs
  • Five-snap front closure
  • Available in white or blue

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size Large/X-Large
Gender Unisex
Material SMS
Color White
Closure Type Snaps
No. of Pockets 3
For Use With (Application) Protects from dust, dirt, grime, and water based liquids
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.