Learn More
Ansell MICROFLEX™ High Five™ Softwear Nitrile Gloves
Catalog No. 19156109 Shop All Ansell Products
Change view
Click to view available options
:
White; Small
Catalog No.
19-156-109 White; Small
1 options
This item is not returnable. View return policy
Catalog No. 19-156-109 Supplier Ansell Supplier No. N791
This item is not returnable. View return policy
Specifications
Certifications/Compliance AQL 1.5, ISO9001 QMS
Color White
Disposable Single-use
Material Nitrile
Powdered Powder Free
Size Small
Thickness (English) 4.5 mil
Thickness (Metric) 0.11 mm
Tensile Strength 20.5
Length (English) 9.5 in.
Length (Metric) 24.13 cm