Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High Five™ Spunbond Polyethylene Shoe Cover

Catalog No. 19156234 Shop All Ansell Products
Change view
Click to view available options
Color: Purple; Size: X-Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-156-234 Color: Purple; Size: X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-156-234 Supplier High Five™ Supplier No. EV0540000B1K
This item is not returnable. View return policy

Specifications

Size X-Large
Material Polyethylene
Color Purple
For Use With (Application) Ideal for protection against water-based contaminants or for general purpose use
Product Type Shoe Covers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.