Learn More
High Five™ Spunbond Polypropylene Coveralls
Catalog No. 19167033 Shop All Ansell Products
Change view
Click to view available options
Cuff Style:
Elastic
Knit
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cuff Style Size
19-167-033 Knit Medium
19-167-914 Elastic 2X-Large
19-167-912 Elastic Large
19-167-913 Elastic X-Large
19-167-915 Elastic 3X-Large
19-167-916 Elastic 4X-Large
19-167-153 Elastic 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-167-033 Supplier High Five™ Supplier No. EW10103M
This item is not returnable. View return policy
Economical protection from dust, dirt and grime
  • Spunbond polypropylene
  • Porous construction for breathability
  • Elastic wrists and ankles
  • Zipper closure
  • White
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Polypropylene
Color White
Cuff Style Knit
Collar Type Standard fold