Learn More
High Five™ Spunbond Polypropylene Shoe Covers with Tread
Catalog No. 19167942 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
X-Large
Includes:
2.5 in. blue and white tread w/ tacky strip
White Tread, 2.5 in. wide tacky strip
Packaging Type:
Bulk Pack
Bulk Vacuum Pack
Dispenser Pack
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes Packaging Type
19-167-942 X-Large 2.5 in. blue and white tread w/ tacky strip Bulk Pack
19-156-236 X-Large White Tread, 2.5 in. wide tacky strip Bulk Vacuum Pack
19-156-235 Universal White Tread, 2.5 in. wide tacky strip Bulk Vacuum Pack
19-167-939 Universal 2.5 in. blue and white tread w/ tacky strip Dispenser Pack
19-167-941 Universal 2.5 in. blue and white tread w/ tacky strip Bulk Pack
19-167-940 X-Large 2.5 in. blue and white tread w/ tacky strip Dispenser Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-167-942 Supplier High Five™ Supplier No. EB1041705B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Sanitary, with 2.5 in. tacky strip on sole to keep wearer from slipping
  • Strong, abrasion-resistant spunbond polypropylene
  • Full elastic top stretches to allow for easy on and off
  • Blue with white tread
Order Info
Bulk pack: 300/case; dispenser pack: three dispensers of 100.
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material Polypropylene
Color Blue
Closure Type Elastic Ankle
Model AS624
Includes 2.5 in. blue and white tread w/ tacky strip
Packaging Quantity 300/Cs.
Packaging Type Bulk Pack
Resistance Abrasion
Product Type Shoe Covers