Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek High Flow Complete Dispense/Waste System
GSA_VA

Catalog No. BT2120532 Shop All Agilent Technologies Inc. Products
Change view
Click to view available options
Capacity (Metric):
4 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
BT2120532 4 L
BT2120534 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BT2120532 Supplier Agilent BioTek Supplier No. 2120532
Add to Cart
Add to Cart
Request a Quote

High Flow Complete Dispense/Waste System for use with the 406 FX Washer Dispenser.

  • Includes four dispense bottles, two waste bottles and high flow vacuum pump

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 4 L
Includes 4 x 4 L Dispense Bottles, 2 x 4 L Waste Bottles, High Flow Vacuum Pump
Type Waste System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.