Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
High molecular weight kininogen (light chain specific) Goat anti-Human, Polyclonal, Nordic

Goat Polyclonal Antibody
Supplier: Vector Laboratories GAHU/HMWK

This item is not returnable.
View return policy
Description
High molecular weight kininogen (light chain specific) Polyclonal antibody specifically detects High molecular weight kininogen (light chain specific) in Human samples. It is validated for ImmunoelectrophoresisSpecifications
High molecular weight kininogen (light chain specific) | |
Polyclonal | |
whole serum with no preservative; pH | |
P01042 | |
Goat | |
1 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoelectrophoresis | |
Unconjugated | |
KNG1 | |
alpha-1-MAP; Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; High molecular weight kininogen; H-kininigen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen; kininogen 1; kininogen 1-like 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; KNG; KNG1; Kng1l1; L-kininogen; low molecular weight (LMW) T-kininogen II; Low molecular weight growth-promoting factor; Lysyl-bradykinin; Major acute phase protein; major acute phase protein (alpha-1-MAP); Thiostatin; T-kinin; T-kininogen 2; T-kininogen 2 heavy chain; T-kininogen 2 light chain; T-kininogen II; T-kininogen II heavy chain; T-kininogen II light chain; Williams-Fitzgerald-Flaujeac factor | |
Human high molecular weight kininogen (light chain specific) | |
RUO | |
3827 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction