Learn More
Restek™ High-Power Electronic Crimping Tool with OLED Display Screen
SDP
Catalog No. p-7199222
Click to view available options
Power Supply:
Power Cord (China)
Power Cord (Europe)
Power Cord (Japan)
Power Cord (Taiwan)
Power Cord (United Kingdom)
Power Cord (United States)
Fixed power supply provides steady, reliable power; Cords are available for China, Europe, Japan, Taiwan, United Kingdom, and United States.
  • Buy only the size crimper or decapper needed (11 mm or 20 mm) with interchangable jaw sets, sold separately
  • The separate crimper and decapper jaw sets fit standard lab vial seals that are aluminum or two-part aluminum/steel caps
  • Ergonomic design reduces strain and arm injury associated with many manual crimping tools
  • Optimize crimping power for each cap type, monitor battery use, and view lifetime statistics with the crisp OLED screen display
  • Save time and minimize adjustments by storing separate programs for different caps and seals
  • Strong enough for use with all steel and magnetic caps
  • Combine with optional base and mounting kit for hands-free operation
  • Two convenient sleep mode options
Specifications
Includes Universal Fixed Power Supply, USB Drive
No. per Pack 1 Ea.
Power Supply Power Cord (China)
Type Crimping Tool
Size 11 or 20 mm
For Use With (Application) Crimp Caps
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.