Learn More
Greiner Bio-One High Profile Polystyrene Microplate Lids
GREENER_CHOICE
Catalog No. p-7075458
Click to view available options
Sterility:
Non-sterile
Sterile
Condensation Rings:
No
Yes
No. per Pack:
1
20
No. per Case:
100
200
For use with non TC-treated 96-well microplates. Greiner Bio-one High Profile Polystyrene Microplate Lids is free of detectable DNase, RNase, human DNA.
  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic, non-cytotoxic
  • Offer protection against contamination and evaporation during sample storage and cell cultivation
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Height (Metric) 9 mm
Condensation Rings Yes
No. per Case 100
No. per Pack 1
For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.