Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Simulation Curriculum Starry Night High School
SCI_ED

Catalog No. s80708 Shop All Simulation Curriculum Products
Click to view available options
Grade:
9 to 12

Starry Night High School features easy-to-use, complete curriculum for high school astronomy education.

  • 35-user annual license
  • 35 Lesson plans joined to exercises and hundreds of simulations, animations & images
  • Teachable Moments feature keeps content fresh and relevant
  • Lessons are correlated to state standards and the NGSS
  • Accurate visualizations allow students to make precise observations of moon phases, motions of the Sun & planets, seasons, eclipses and much more
  • Interactive exercises and hands-on activities encourage questioning, experimentation and exploration and accommodate diverse learning styles
  • Pre- and post-assessment resources with recommendations for improved student performance
  • Flexible teaching models for easy adaptation to pacing and classroom needs
  • Opportunities to develop key skills: observation, data collection, analytical thinking and discovery
  • Extensions that connect to topics in math, physics, language arts, social studies and technology
  • Access for the Entire Classroom, Teachers and Staff
  • Includes digital teacher's guide with helpful how-to's and support resources
  • Runs in the browser using a standard Internet connection
  • Chromebook™ iPad™ Android™ Windows™ and Mac™ Compliant

Order Info

For Grades 9-12.

Specifications

Grade 9 to 12
For Use With (Application) Astronomy education
Product Type Educational software
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.