Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ High Select™ HSA/Immunoglobulin Depletion Resin, 50 mL PROMO

Catalog No. PIA36368PM
Click to view available options
:
Thermo Scientific™ High Select™ HSA/Immunoglobulin Depletion Resin

Decrease albumin, immunoglobulins, and other high-abundance proteins from human serum or plasma in preparation for mass spectrometry analysis, or 1D and 2D gel electrophoresis using High Select depletion spin columns or resin.

  • Flexible—resin provided in mini and midi column formats as well as bulk for custom filling options
  • Optimized—removes >95% of IgG and >95% of albumin (HSA/Immunoglobulin); removes >95% of IgG and >95% of albumin, plus 12 other abundant proteins (Top 14)
  • Fast—process samples in 10 minutes
  • Economical—cost-effective spin columns are priced for single use
  • Reduced carry-over—one-time use prevents protein carryover and experimental variability

High Select HSA/Immunoglobulin Depletion Resin uses highly specific, immobilized anti-HSA and anti-immuoglobulin antibodies (IgG, IgA, IgM, IgD, and IgE) to remove human serum albumin (HSA) and all major subclasses of immunoglobulins from serum and plasma. High Select Top14 Abundant Protein Depletion Resin uses highly specific immobilized antibodies to remove human serum albumin (HSA), albumin, IgG, IgA, IgM, IgD, IgE, kappa and lambda light chains, alpha-1-acidglycoprotein, alpha-1-antitrypsin, alpha-2-macroglobulin, apolipoprotein A1, fibrinogen, haptoglobin, and transferrin from serum, plasma, or spinal fluids. The resins are provided in an economical and convenient spin column format specifically designed for one-step processing and single use or bulk format for custom filling.

Specifications

For Use With (Equipment) Mass Spectrometer
Product Type Resin
Product Line High Select™
Content And Storage Store at 2°C to 8°C
Label or Dye Unlabeled
Shipping Condition Wet Ice
Starting Material Plasma, Serum
Final Product Type Protein
Workflow Step Protein Depletion
Quantity 50 mL
Purification Target HSA/Immunoglobulin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.