Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedical™ High Sensitivity C-Reactive Protein (hsCRP) ELISA Kit
Click to view available options
Quantity:
96 Wells
Description
The High Sensitivity C-Reactive Protein (hsCRP) ELISA Kit is intended for the quantitative determination of C-reactive protein in human serum. Enhanced sensitivity measurements of CRP can be useful for the detection and evaluation of infection, tissue injury, inflammatory disorders and associated diseases.

Specifications
Specifications
Product Type | Kit and Assay |
Sample Type | Serum |
Target Species | Human |
For Use With (Application) | Endocrinology |
Quantity | 96 Wells |
Regulatory Status | FDA Cleared |
Sample Volume | 5 μL |
Storage Requirements | 2°C to 8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction