Learn More
Restek™ High Temperature Electron Ionization (EI) Filament
SDP
Catalog No. 06611986 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Equipment):
Agilent 5972/5973/5975/5977 GC-MS
Catalog No. For Use With (Equipment)
06-611-986 Agilent 5972/5973/5975/5977 GC-MS
1 options
Catalog No. 06-611-986 Supplier Restek™ Supplier No. 23099
For Agilent 5972, 5973, 5975 or 5977 GC-MS
  • Filaments meet or exceed original manufacturer's performance
  • Every filament is subjected to QC tests including heat, electrical current and resistance
  • Samples from each filament manufacturing lot are installed in MSD for in situ testing
Specifications
Type Hight Temperature Electron Ionization Filament
For Use With (Equipment) Agilent 5972/5973/5975/5977 GC-MS