Learn More
R&D Systems™ High-Throughput Glutathione Peroxidase Assay Kit
SDP
Catalog No. 7512100K Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
480 Tests
Catalog No. Quantity
75-121-00K 480 Tests
1 options
This item is not returnable. View return policy
Catalog No. 75-121-00K Supplier R&D Systems™ Supplier No. 7512100K
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Includes
Glutathione Peroxidase, GSH and NADPH, Glutathione Reductase, Cumene Hydroperoxide, Buffer, 5 x 96-well Plates
Spectrophotometric assay for the quantitation of glutathione peroxidase in cells, erythrocytes, blood, and tissues
Glutathione peroxidase is a tetramer of four identical subunits with MW 84,000. It is found in mammalian cells and helps prevent lipid peroxidation of cell membranes by consuming peroxide in the cell.
Sufficient reagents for 480 Tests-96 well Format or 100 Tests-cuvette Format
Rate of decrease in the A340 is directly proportional to the glutathione peroxidase activity in the sample
Specifications
Content And Storage The product requires storage at -20 to -70° C and room temperature. Consult the product insert for specific storage temperatures. Do not use past expiration date.
Quantity 480 Tests
Product Line R&D Systems