Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIKESHI Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139744
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5139744 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5139744 Supplier Invitrogen™ Supplier No. PA5139744
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: LGFVTNGKPS AIFKISGLKS GEGSQHPFGA MNIVRTPSVA QIGISVELLD SMAQQTPVGN AAVSSVDSFT QFTQKMLDNF YNFASSFAVS QAQMTPSPSE MFIPANVVLK WYENFQRRLA QN Highest antigen sequence indentity to the following orthologs: Mouse - 97%, Rat - 97%.

Required for organization and/or function of the secretory apparatus in Clara cells in lung.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIKESHI
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene HIKESHI
Gene Accession No. Q53FT3
Gene Alias 0610007P06Rik; 1110002N09Rik; C11orf73; C1H11orf73; C21H11orf73; C29H11orf73; chromosome unknown open reading frame, human C11orf73; heat shock protein nuclear import factor hikeshi; Hikeshi; Hikeshi, heat shock protein nuclear import factor; HLD13; HSPC138; HSPC179; HSPC248; hypothetical protein LOC504867; hypothetical protein LOC550239; l(7)6Rn; l7Rn6; lethal gene on chromosome 7 Rinchik 6 protein; lethal, Chr 7, Rinchik 6; LOC100762248; OPI10; protein Hikeshi; Protein of unknown function DUF775 containing protein; similar to hypothetical protein HSPC138; uncharacterized protein C11orf73 homolog; Uncharacterized protein C11orf73-like; unknown; zgc:110091
Gene Symbols HIKESHI
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HIKESHI. Recombinant protein control fragment (Product #RP-108721).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51501
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.