Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HIN-1 Monoclonal Antibody (01)
GREENER_CHOICE

Catalog No. PIMA531160
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531160 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531160 Supplier Invitrogen™ Supplier No. MA531160
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human SCGB3A1/PnSP-2.

HIN-1, also known as High In Normal-1 or Secretoglobin Family 3A Member 1 (SCGB3A1), is a gene located on chromosome 5q33.3. It encodes a protein that is predominantly expressed in normal mammary epithelial cells and is thought to function in epithelial cell signaling, contributing to cellular differentiation and maintaining normal tissue architecture. The HIN-1 gene is often silenced or downregulated in various cancers, particularly breast cancer, due to hypermethylation of its promoter region, suggesting a potential tumor suppressor role. Loss of HIN-1 expression has been implicated in the progression of malignancies by disrupting normal cell signaling pathways, thereby facilitating oncogenesis. Given its significant downregulation in cancerous tissues, HIN-1 is being explored as a biomarker for early cancer detection and may provide insights into novel therapeutic strategies that aim to restore its expression and normal cellular functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIN-1
Applications ELISA
Classification Monoclonal
Clone 01
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SCGB3A1
Gene Accession No. Q96QR1
Gene Alias cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2
Gene Symbols SCGB3A1
Host Species Mouse
Immunogen Recombinant Human SCGB3A1/PnSP-2 Protein (Met1-Gly104).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92304
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.