Learn More
Invitrogen™ HIN-1 Monoclonal Antibody (3G5)

Description
Peptide Sequence: MKLAALLGLC VALSCSSAAA FLVGSAKPVA QPVAALESAA EAGAGTLANP LGTLNPLKLL LSSLGIPVNH LIEGSQKCVA ELGPQAVGAV KALKALLGAL TVFG.
Specifications
Specifications
| Antigen | HIN-1 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 3G5 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | SCGB3A1 |
| Gene Accession No. | Q96QR1 |
| Gene Alias | cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.