Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIN-1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5121911

This item is not returnable.
View return policy
Description
Potential growth inhibitory cytokine. Classified as a secretoglobin; may have a role in airway function.
Specifications
HIN-1 | |
Polyclonal | |
Unconjugated | |
SCGB3A1 | |
cytokine high in normal-1; cytokine HIN-1; High in normal 1; HIN1; HIN-1; LU105; pneumo secretory protein 2; PNSP2; PnSP-2; SCGB3A1; Secretoglobin family 3A member 1; secretoglobin, family 3A, member 1; UGRP2; UGRP2MGC87867; UNQ629/PRO1245; Uteroglobin-related protein 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100515999, 92304 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q96QR1 | |
SCGB3A1 | |
Phe21-Gly104 (Accession Q96QR1), with N-terminal His and GST Tag | |
100 μL | |
Primary | |
Human, Pig | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction