Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HindIII (10 U/μL)

The HindIII restriction enzyme recognizes A^AGCTT sites and cuts best at 37°C in R buffer.

$55.80 - $223.50

Specifications

Concentration 10 U/μL
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
FERER0502
View Documents
Thermo Scientific™
ER0502
5 x 5,000 units
Each for $223.50
Only null left
Add to Cart
 
FERER0505
View Documents
Thermo Scientific™
ER0505
10,000 units
Each for $115.00
Only null left
Add to Cart
 
FERER0501
View Documents
Thermo Scientific™
ER0501
5,000 units
Each for $55.80
Only null left
Edge
Add to Cart
 
Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality – stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.