Learn More
Azer Scientific Hinged Biopsy Cassette, Taped
Description
To be used with Leica and Sakura Ink Jet printers. Molded from acetal, they keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories.
The biopsy cassette is designed to hold small samples securely during processing. The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface.
The hinged biopsy cassette incorporates a unique recessed cover, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets. The cassette ensures efficient fluid exchange and drainage thanks to 114 openings each measuring 1 x 5 mm. The anterior writing area is slanted at a 45° angle.
Features:
- Taped cassettes to be used with Leica and Sakura Ink Jet Printers
- Recessed cover, a great space saving feature
- Easy-to-remove pre-attached disposable cover
- Will resist most commonly used histological solvents
- 1 mm square openings to maximize fluid exchange
Specifications
Specifications
| Closure Included | Yes |
| Closure Type | Lid |
| Color | Yellow |
| Product Type | Biopsy Cassette |