Learn More
Azer Scientific Hinged Biopsy Cassette, Taped
SDP
Catalog No. p-200025599
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Biopsy Cassette, taped for automation, lids hinged at front, back tab, 45° writing angle, 40/stack, 50 stacks/case.
To be used with Leica and Sakura Ink Jet printers. Molded from acetal, they keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories.
The biopsy cassette is designed to hold small samples securely during processing. The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface.
The hinged biopsy cassette incorporates a unique recessed cover, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets. The cassette ensures efficient fluid exchange and drainage thanks to 114 openings each measuring 1 x 5 mm. The anterior writing area is slanted at a 45° angle.
Features:
  • Taped cassettes to be used with Leica and Sakura Ink Jet Printers
  • Recessed cover, a great space saving feature
  • Easy-to-remove pre-attached disposable cover
  • Will resist most commonly used histological solvents
  • 1 mm square openings to maximize fluid exchange
Specifications
Closure Included Yes
Closure Type Lid
Color Aqua
Product Type Biopsy Cassette
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.