Learn More
Azer Scientific Hinged Mesh Cassette,Taped
Description
This hinged cassette offers 1676 square openings (0.38 mm) allowing for a greatly improved fluid exchange and drainage without having to use biopsy pads. Large anterior slots in both cassette and cover ensure that the hinged biopsy cassette will sink rapidly. A large square compartment measuring 27 mm is perfect even for needle biopsies. The cover does not protrude above the cassette, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and tissue processors.
Molded from acetal, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. Anterior writing area is at a 45° angle to make the cassette more suitable to be used with automated cassette printers.
Features:
- No biopsy pad necessary
- Recessed cover, a great space saving feature
- Easy-to-remove pre-attached disposable cover
- Taped cassettes to be used with Leica and Sakura Ink Jet Printers
Specifications
Specifications
| Closure Included | Yes |
| Closure Type | Lid |
| Color | Aqua |
| Product Type | Biopsy Cassette |