Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Hinged Tissue Cassette, Taped
SDP

Catalog No. 22026786
Change view
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-026-786 Aqua
22-026-787 Blue
22-026-788 Green
22-026-790 Gray
22-026-791 Lilac
22-026-792 Orange
22-026-793 Pink
22-026-794 Peach
22-026-795 Tan
22-026-796 White
22-026-797 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
This item is not returnable. View return policy
Catalog No. 22-026-786 Supplier Azer Scientific Supplier No. ES6015A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Tissue Cassette, taped for automation, lids hinged at front, back tab, 45° writing angle, 40/stack, 50 stacks/case.

To be used with Leica and Sakura Ink Jet printers. Molded from acetal, they keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories.

The hinged tissue cassette incorporates a unique recessed cover, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets. The cassette ensures efficient fluid exchange and drainage thanks to 114 openings each measuring 1 x 5 mm. The anterior writing area is slanted at a 45° angle.

Features:

  • Taped cassettes to be used with Leica and Sakura Ink Jet Printers
  • Recessed cover, a great space saving feature
  • Easy-to-remove pre-attached disposable cover

Specifications

Closure Included Yes
Closure Type Lid
Color Aqua
Product Type Tissue Cassette
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.