Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azer Scientific Hinged Tissue Cassette, Taped
SDP

Catalog No. p-200025598
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Tissue Cassette, taped for automation, lids hinged at front, back tab, 45° writing angle, 40/stack, 50 stacks/case.

To be used with Leica and Sakura Ink Jet printers. Molded from acetal, they keep specimens safely submerged and are resistant to the chemical action of most solvents used in histology laboratories.

The hinged tissue cassette incorporates a unique recessed cover, a great space saving feature allowing more cassettes to be stacked in automatic labeling machines and in storage cabinets. The cassette ensures efficient fluid exchange and drainage thanks to 114 openings each measuring 1 x 5 mm. The anterior writing area is slanted at a 45° angle.

Features:

  • Taped cassettes to be used with Leica and Sakura Ink Jet Printers
  • Recessed cover, a great space saving feature
  • Easy-to-remove pre-attached disposable cover

Specifications

Closure Included Yes
Closure Type Lid
Color Aqua
Product Type Tissue Cassette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.