Learn More
Invitrogen™ HINT1 Polyclonal Antibody

Description
A suggested positive control is Jurkat cell lysate. PA5-20613 can be used with blocking peptide PEP-0733.
Specifications
Specifications
| Antigen | HINT1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Hint1 |
| Gene Accession No. | P49773, P62959, P70349 |
| Gene Alias | 17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1 |
| Gene Symbols | Hint1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.