Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HINT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560760
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560760 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560760 Supplier Invitrogen™ Supplier No. PA560760
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IPKKHISQIS VAEDDDESLL GHLMIVGKKC AADLGLNKGY RMVVNEGSDG GQSVYHVHLH VL Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

Histidine triad nucleotide-binding protein 1 (HINT1) is a member of the histidine triad (HIT) protein family, a group of small nucleotide-binding and -hydrolyzing proteins. HINT1 interacts with several diverse proteins and has been suggested to have tumor suppressive activities. HINT1 catalyzes the hydrolysis of adenosine 5'-monophoramidate substrates such as AMP-morpholidate, but its enzymatic function does not appear to play a role in its tumor suppression. Recent experiments demonstrate that HINT1 forms a complex with POSH and JNK in vivo, inhibiting AP-1 activity and the phosphorylation of c-Jun, and this action could contribute to the tumor suppressor activity of HINT1. Other studies raise the possibility of HINT1 as a candidate gene for schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HINT1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Hint1
Gene Accession No. P49773, P62959, P70349
Gene Alias 17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1
Gene Symbols Hint1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human HINT1. Recombinant protein control fragment (Product #RP-96928).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15254, 3094, 690660
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.