Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ HINT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA579363
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA579363 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA579363 Supplier Invitrogen™ Supplier No. PA579363
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human Jurkat whole cell, human Raji whole cell, human Hela whole cell, rat brain tissue, rat thymus tissue, mouse brain tissue, mouse thymus tissue. IHC: mouse intestine tissue, rat kidney tissue, human intestinal cancer tissue. ICC/IF: A431 cell. Flow: A549 cell.

Histidine triad nucleotide-binding protein 1 (HINT1) is a member of the histidine triad (HIT) protein family, a group of small nucleotide-binding and -hydrolyzing proteins. HINT1 interacts with several diverse proteins and has been suggested to have tumor suppressive activities. HINT1 catalyzes the hydrolysis of adenosine 5'-monophoramidate substrates such as AMP-morpholidate, but its enzymatic function does not appear to play a role in its tumor suppression. Recent experiments demonstrate that HINT1 forms a complex with POSH and JNK in vivo, inhibiting AP-1 activity and the phosphorylation of c-Jun, and this action could contribute to the tumor suppressor activity of HINT1. Other studies raise the possibility of HINT1 as a candidate gene for schizophrenia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HINT1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Hint1
Gene Accession No. P49773, P62959, P70349
Gene Alias 17 kDa inhibitor of protein kinase C; AA673479; Adenosine 5'-monophosphoramidase; FLJ30414; FLJ32340; HINT; Hint1; histidine triad nucleotide binding protein 1; histidine triad nucleotide-binding protein 1; Ipk1; NMAN; PKC inhibitor/ interacting protein; Pkci; PKCI1; PKCI-1; Prkci; PRKCNH1; protein kinase C inhibitor 1; protein kinase C, iota; protein kinase C-interacting protein 1
Gene Symbols Hint1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence in the middle region of human HINT1 (59-92aa HISQISVAEDDDESLLGHLMIVGKKCAADLGLNK).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 15254, 3094, 690660
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.