Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HINT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | HINT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
HINT2 Polyclonal specifically detects HINT2 in Human samples. It is validated for Western Blot.Specifications
HINT2 | |
Polyclonal | |
Rabbit | |
NP_115982 | |
84681 | |
Synthetic peptide directed towards the C terminal of human HINT2The immunogen for this antibody is HINT2. Peptide sequence RISQAEEEDQQLLGHLLLVAKQTAKAEGLGDGYRLVINDGKLGAQSVYHL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
HINT2 | |
IgG | |
17 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title