Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HINT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117985
This item is not returnable.
View return policy
Description
HINT3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Histidine triad proteins, such as HINT3, are nucleotide hydrolases and transferases that act on the alpha-phosphate of ribonucleotides. Hydrolyzes phosphoramidate and acyl-adenylate substrates.
Specifications
HINT3 | |
Polyclonal | |
PBS with 0.03% ProClin 300; pH 7 | |
Q9NQE9 | |
HINT3 | |
E. coli-derived Human HINT3 fragment. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
Unconjugated | |
HINT3 | |
HINT3; HINT-3; HINT4; histidine triad nucleotide binding protein 3; histidine triad nucleotide-binding protein 3; HIT-like protein | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
135114 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction