Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIP-55 Antibody (OTI6C9), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271841G
Description
HIP-55 Monoclonal specifically detects HIP-55 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
| HIP-55 | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DBNL | |
| Full length recombinant protein of human HIP-55 (NP_054782) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Immunohistochemistry | |
| OTI6C9 | |
| Immunohistochemistry | |
| ABP1, Cervical SH3P7, CMAP, Drebrin-F, drebrin-like, drebrin-like protein, HIP-55SH3P7Cervical mucin-associated protein, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, src homology 3 domain-containing protein HIP-55 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 28988.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction