Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hip Antibody (HL1737) - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP325505
Description
Hip Monoclonal antibody specifically detects Hip in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Hip | |
Monoclonal | |
Unconjugated | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
64399 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HL1737 | |
Western Blot 1:500-1:3000, Immunohistochemistry, Immunohistochemistry-Paraffin | |
FLJ90230, hedgehog interacting protein, hedgehog-interacting protein, HIPFLJ20992 | |
Recombinant protein encompassing a sequence within the center region of human Hip. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction