Learn More
3B Scientific™ Hip, Knee and Elbow Joint Models - includes 3B Smart Anatomy


Shop All 3B Scientific Products



Description
3B Smart Anatomy is now included with any 3B Scientific anatomy model.
Every original 3B Scientific anatomy model now includes these additional FREE features:
- Free access to the anatomy course 3B Smart Anatomy, hosted inside the award-winning Complete Anatomy app by 3D4Medical
- The 3B Smart Anatomy course includes 23 digital anatomy lectures, 117 different virtual anatomy models and 39 anatomy quizzes to test your knowledge
- FREE warranty upgrade from 3 to 5 years with every product registration
TIPP: you will also receive access to a free 3-day trial to all premium features of the Complete Anatomy app when you sign up for your 3B Smart Anatomy course.
To unlock these benefits, simply scan the label located on your model and register online. All 3B Smart Anatomy features are completely free of charge.
These models represent clearly the relationship between the muscular and skeletal systems.
- Muscles of leg are easily removable to permit study of deeper anatomical layers
- Color-coded and raised areas help in differentiation
- Muscles can be attached to and removed from the corresponding areas of origin and insertion
Specifications
Specifications
Product Type | Model, Hip Joint with Removable Muscles |
For Use With (Application) | For educational purposes |
Includes | Removable muscles |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.