Learn More
HIP Monoclonal Antibody (2G6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA3413
Description
MA3-413 detects heat shock cognate protein 70 kDa (HSC70)-interacting protein (Hip) from human, mouse, chicken and rabbit samples. MA3-413 has been successfully used in Western blot, immunofluorescence, and immunoprecipitation procedures. By Western blot, this antibody detects a 48 kDa protein representing Hip from rabbit reticulocyte lysate. The MA3-413 antigen is recombinant human Hip protein.
The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene.
            Specifications
| HIP | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites diluted in PBS with 0.05% sodium azide | |
| P50502, Q5ZLF0, Q99L47 | |
| St13 | |
| Recombinant human Hip protein. | |
| RUO | |
| 100354640, 418003, 6767, 70356 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid | 
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 2G6 | |
| Unconjugated | |
| St13 | |
| 1110007I03Rik; 3110002K08Rik; AAG2; aging-associated protein 2; AW555194; FAM10A1; FAM10A4; heat shock 70kD protein binding protein; hedgehog interacting protein; hedgehog-interacting protein; HHIP; HIP; HOP; Hsc70-interacting protein; Hsp70 interacting protein; Hsp70-interacting protein; HSPABP; HSPABP1; hypothetical protein; p48; PRO0786; Progesterone receptor-associated p48 protein; Protein FAM10A1; Protein ST13 homolog; putative tumor suppressor ST13; RCJMB04_6h13; Renal carcinoma antigen NY-REN-33; SNC6; ST13; ST13 Hsp70 interacting protein; ST13, Hsp70 interacting protein; ST13P5; suppression of tumorigenicity 13; suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein); suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5; suppression of tumorigenicity 13 (colon carcinoma) Hsp70-interacting protein; suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m; UNQ5825/PRO19644 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Chicken, Human, Mouse, Rabbit | |
| Antibody | |
| IgG | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.