Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HIP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583818
Description
Immunogen sequence: PGMAGMPGLN EILSDPEVLA AMQDPEVMVA FQDVAQNPAN MSKYQSNPKV MNLISKLSAK FGGQA.
The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene.
Specifications
HIP | |
Polyclonal | |
Unconjugated | |
St13 | |
1110007I03Rik; 3110002K08Rik; AAG2; aging-associated protein 2; AW555194; FAM10A1; FAM10A4; heat shock 70kD protein binding protein; hedgehog interacting protein; hedgehog-interacting protein; HHIP; HIP; HOP; Hsc70-interacting protein; Hsp70 interacting protein; Hsp70-interacting protein; HSPABP; HSPABP1; hypothetical protein; p48; PRO0786; Progesterone receptor-associated p48 protein; Protein FAM10A1; Protein ST13 homolog; putative tumor suppressor ST13; RCJMB04_6h13; Renal carcinoma antigen NY-REN-33; SNC6; ST13; ST13 Hsp70 interacting protein; ST13, Hsp70 interacting protein; ST13P5; suppression of tumorigenicity 13; suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein); suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5; suppression of tumorigenicity 13 (colon carcinoma) Hsp70-interacting protein; suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m; UNQ5825/PRO19644 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6767 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P50502 | |
St13 | |
Recombinant protein corresponding to Human ST13. Recombinant protein control fragment (Product #RP-99512). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction